siRNA Details
| Gene Symbol: Gm31462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136698.1 | - | 1 | 121 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721090.1 | - | 1 | 121 | Chr. 15 - 81005407 - 81006410 [-] on Build GRCm38 | |

Gm31462
Genomic Map