siRNA Details
| Gene Symbol: Gm5095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033454.1 | 1 | 214 | |||
| GenBank mRNA | AK043399.1 | - | 1 | 215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5095| Gene Symbol: Gm5095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033454.1 | 1 | 214 | |||
| GenBank mRNA | AK043399.1 | - | 1 | 215 | ||