siRNA Details
| Gene Symbol: 1700001J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019587.1 | - | 1 | 760 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF104643.1 | - | 2 | 506 | Chr. 11 - 114185973 - 114186674 [-] on Build GRCm38 | |

1700001J04Rik
Genomic Map