siRNA Details
| Gene Symbol: 4930428D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019586.1 | - | 1 | 60 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714774.1 | - | 1 | 60 | Chr. 16 - 69796140 - 69800381 [-] on Build GRCm38 | |

4930428D20Rik
Genomic Map