siRNA Details
| Gene Symbol: 4930425L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015569.1 | - | 266 | |||
| AK019585.1 | - | 4 | 645 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930425L21Rik| Gene Symbol: 4930425L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015569.1 | - | 266 | |||
| AK019585.1 | - | 4 | 645 | |||