siRNA Details
Gene Symbol: Gm16063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046178.1 | 4 | 1300 | |||
GenBank mRNA | AK090161.1 | - | 4 | 1300 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046178.1 | 4 | 1300 | |||
GenBank mRNA | AK090161.1 | - | 4 | 1300 |