siRNA Details
| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080684.1 | - | 3 | 1760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4c| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080684.1 | - | 3 | 1760 | ||