siRNA Details
| Gene Symbol: 2900027M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013604.1 | - | 1 | 1700 | ||
| AK053575.1 | - | 1 | 114 | |||
| BB665194.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900027M19Rik| Gene Symbol: 2900027M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013604.1 | - | 1 | 1700 | ||
| AK053575.1 | - | 1 | 114 | |||
| BB665194.1 | - | 1 | 114 | |||