siRNA Details
| Gene Symbol: Ilf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052536.1 | - | 1 | 2547 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ilf3| Gene Symbol: Ilf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052536.1 | - | 1 | 2547 | ||