siRNA Details
| Gene Symbol: 4930555G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016129.1 | - | 1 | 29 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715732.1 | - | 1 | 29 | Chr. 10 - 87230760 - 87239248 [+] on Build GRCm38 | |

4930555G07Rik
Genomic Map