siRNA Details
| Gene Symbol: 4933404I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016650.1 | - | 2 | 634 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716337.1 | - | 2 | 634 | Chr. 7 - 40262705 - 40269507 [+] on Build GRCm38 | |

4933404I11Rik
Genomic Map