siRNA Details
| Gene Symbol: D530043N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021315.1 | - | 1 | 204 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730676.1 | - | 1 | 201 | Chr. 2 - 113650466 - 113650922 [+] on Build GRCm38 | |

D530043N20Rik
Genomic Map