siRNA Details
| Gene Symbol: 1700092C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045467.1 | 3 | 336 | |||
| GenBank mRNA | AK007037.1 | - | 3 | 337 | ||
| AK018991.1 | - | 3 | 337 | |||
| BY707399.1 | - | 3 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700092C02Rik| Gene Symbol: 1700092C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045467.1 | 3 | 336 | |||
| GenBank mRNA | AK007037.1 | - | 3 | 337 | ||
| AK018991.1 | - | 3 | 337 | |||
| BY707399.1 | - | 3 | 337 | |||