siRNA Details
| Gene Symbol: 1700092C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045467.1 | 1 | 97 | |||
| GenBank mRNA | AK007037.1 | - | 1 | 98 | ||
| AK018991.1 | - | 1 | 98 | |||
| BY707399.1 | - | 1 | 98 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707105.1 | - | 1 | 105 | Chr. 8 - 77624172 - 77628954 [+] on Build GRCm38 | |

1700092C02Rik
Genomic Map