siRNA Details
| Gene Symbol: 1700013N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005966.1 | - | 140 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706098.1 | - | 3 | 140 | Chr. 12 - 106836868 - 106877558 [+] on Build GRCm38 | |

1700013N06Rik
Genomic Map