siRNA Details
| Gene Symbol: 1700013N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005966.1 | - | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700013N06Rik| Gene Symbol: 1700013N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005966.1 | - | 539 | |||