siRNA Details
Gene Symbol: 1700013K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005961.1 | - | 1 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706093.1 | - | 1 | 118 | Chr. 9 - 62267376 - 62267843 [-] on Build GRCm38 |
