siRNA Details
| Gene Symbol: 1700013K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005961.1 | - | 1 | 401 | ||
| AV040749.2 | - | 1 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700013K18Rik| Gene Symbol: 1700013K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005961.1 | - | 1 | 401 | ||
| AV040749.2 | - | 1 | 226 | |||