siRNA Details
| Gene Symbol: 2210019I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038157.1 | 1, 2 | 309 | |||
| GenBank mRNA | AK008755.1 | - | 1, 2 | 309 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708657.1 | - | 1, 2 | 306 | Chr. 5 - 147229027 - 147271519 [-] on Build GRCm38 | |

2210019I11Rik
Genomic Map