siRNA Details
Gene Symbol: 2210019I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038157.1 | 6 | 942 | |||
GenBank mRNA | AK008755.1 | - | 6 | 942 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2210019I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038157.1 | 6 | 942 | |||
GenBank mRNA | AK008755.1 | - | 6 | 942 |