siRNA Details
| Gene Symbol: 9330154F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020373.1 | - | 1 | 48 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721342.1 | - | 1 | 45 | Chr. 4 - 68800982 - 68801628 [-] on Build GRCm38 | |

9330154F10Rik
Genomic Map