siRNA Details
| Gene Symbol: Gm36617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076851.1 | - | 4 | 476 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV265592.2 | - | 4 | 440 | Chr. 15 - 59275431 - 59291946 [+] on Build GRCm38 | |

Gm36617
Genomic Map