siRNA Details
| Gene Symbol: 8030423J24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020194.1 | - | 3 | 353 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720413.1 | - | 3 | 353 | Chr. 13 - 70882948 - 70884306 [+] on Build GRCm38 | |

8030423J24Rik
Genomic Map