siRNA Details
| Gene Symbol: 8030442B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040615.1 | 3 | 484 | |||
| GenBank mRNA | AK020198.1 | - | 3 | 485 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720418.1 | - | 3 | 485 | Chr. 2 - 11338758 - 11398500 [-] on Build GRCm38 | |

8030442B05Rik
Genomic Map