siRNA Details
| Gene Symbol: 8030442B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040615.1 | 3 | 847 | |||
| GenBank mRNA | AK020198.1 | - | 3 | 848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8030442B05Rik| Gene Symbol: 8030442B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040615.1 | 3 | 847 | |||
| GenBank mRNA | AK020198.1 | - | 3 | 848 | ||