siRNA Details
| Gene Symbol: Ggps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF014450.1 | - | 1 | 402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggps1| Gene Symbol: Ggps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF014450.1 | - | 1 | 402 | ||