siRNA Details
| Gene Symbol: 1700008K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038141.1 | 2 | 886 | |||
| NR_038142.1 | 3 | 789 | ||||
| NR_038143.1 | 3 | 795 | ||||
| NR_038144.1 | 2 | 742 | ||||
| NR_038145.1 | 2 | 625 | ||||
| GenBank mRNA | AK005780.1 | - | 3 | 772 | ||

1700008K24Rik
Genomic Map