siRNA Details
| Gene Symbol: Gm39888 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040511.1 | - | 2 | 347 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB590223.2 | - | 2 | 343 | Chr. 2 - 101991980 - 102009016 [+] on Build GRCm38 | |

Gm39888
Genomic Map