siRNA Details
| Gene Symbol: Gm10791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045889.1 | 4 | 1204 | |||
| GenBank mRNA | AK082307.1 | - | 3 | 2980 | ||
| AK166404.1 | - | 4 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10791| Gene Symbol: Gm10791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045889.1 | 4 | 1204 | |||
| GenBank mRNA | AK082307.1 | - | 3 | 2980 | ||
| AK166404.1 | - | 4 | 786 | |||