siRNA Details
| Gene Symbol: Gm2109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046066.1 | 3 | 533 | |||
| GenBank mRNA | AK082308.1 | - | 3 | 534 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650052.1 | - | 3 | 534 | Chr. 9 - 83691475 - 83692452 [+] on Build GRCm38 | |

Gm2109
Genomic Map