siRNA Details
| Gene Symbol: Gm2109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046066.1 | 3 | 1232 | |||
| GenBank mRNA | AK082308.1 | - | 3 | 1233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2109| Gene Symbol: Gm2109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046066.1 | 3 | 1232 | |||
| GenBank mRNA | AK082308.1 | - | 3 | 1233 | ||