siRNA Details
| Gene Symbol: 1810056I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019035.1 | - | 1 | 145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810056I18Rik| Gene Symbol: 1810056I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019035.1 | - | 1 | 145 | ||