siRNA Details
| Gene Symbol: Gm15997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045423.1 | 5 | 1444 | |||
| GenBank mRNA | AK043895.1 | - | 5 | 1445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15997| Gene Symbol: Gm15997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045423.1 | 5 | 1444 | |||
| GenBank mRNA | AK043895.1 | - | 5 | 1445 | ||