siRNA Details
| Gene Symbol: Pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW048905.1 | - | 1 | 64 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK014830.1 | - | 632 | Chr. - - [-] on Build | ||

Pacsin3
Genomic Map