siRNA Details
| Gene Symbol: Scarletltr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040743.1 | 2, 3 | 190 | |||
| GenBank mRNA | AK011166.1 | - | 2, 3 | 191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710499.1 | - | 2, 3 | 191 | Chr. 9 - 64080761 - 64088046 [+] on Build GRCm38 | |

Scarletltr
Genomic Map