siRNA Details
| Gene Symbol: 1190001M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004435.1 | - | 1 | 200 | ||
| BC023846.1 | - | 1 | 939 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704663.1 | - | 1 | 197 | Chr. 11 - 82974316 - 82974854 [-] on Build GRCm38 | |

1190001M18Rik
Genomic Map