siRNA Details
| Gene Symbol: E130002L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040777.1 | - | 2 | 2418 | ||
| AK053256.1 | - | 1 | 2948 | |||
| BB215043.2 | - | 1 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130002L11Rik| Gene Symbol: E130002L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040777.1 | - | 2 | 2418 | ||
| AK053256.1 | - | 1 | 2948 | |||
| BB215043.2 | - | 1 | 392 | |||