siRNA Details
| Gene Symbol: E130002L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040777.1 | - | 2 | 414 | ||
| AK053256.1 | - | 1 | 944 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB636708.1 | - | 2 | 414 | Chr. 12 - 78959030 - 78960214 [-] on Build GRCm38 | |

E130002L11Rik
Genomic Map