siRNA Details
| Gene Symbol: 9230104K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020313.1 | - | 1 | 320 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI696898.1 | - | 1 | 46 | Chr. 4 - 45406256 - 45406914 [-] on Build GRCm38 | |

9230104K21Rik
Genomic Map