siRNA Details
| Gene Symbol: A730018C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036459.1 | 1 | 24 | |||
| GenBank mRNA | AK042713.1 | - | 1 | 38 | ||
| AK048513.1 | - | 1 | 25 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB649310.1 | - | 1 | 23 | Chr. 12 - 112412161 - 112423197 [-] on Build GRCm38 | |

A730018C14Rik
Genomic Map