siRNA Details
| Gene Symbol: Gm32461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085322.1 | - | 3 | 287 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB660906.1 | - | 3 | 287 | Chr. 16 - 86770379 - 86806513 [-] on Build GRCm38 | |

Gm32461
Genomic Map