siRNA Details
| Gene Symbol: Ccr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038272.1 | - | 1 | 3497 | ||
| AK162996.1 | - | 1 | 3638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr9| Gene Symbol: Ccr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038272.1 | - | 1 | 3497 | ||
| AK162996.1 | - | 1 | 3638 | |||