siRNA Details
| Gene Symbol: Usp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083494.1 | - | 1 | 2925 | ||
| BC082334.1 | - | 3 | 4266 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB442079.2 | - | 1 | 565 | Chr. 7 - 6769493 - 6770139 [+] on Build GRCm38 | |

Usp29
Genomic Map