siRNA Details
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF227912.1 | - | 4 | 1741 | ||
AK083494.1 | - | 1 | 28 | |||
BC082334.1 | - | 3 | 1374 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF227912.1 | - | 4 | 1741 | ||
AK083494.1 | - | 1 | 28 | |||
BC082334.1 | - | 3 | 1374 |