siRNA Details
| Gene Symbol: Usp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF227912.1 | - | 4 | 1767 | ||
| AK083494.1 | - | 1 | 54 | |||
| BC082334.1 | - | 3 | 1400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp29| Gene Symbol: Usp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF227912.1 | - | 4 | 1767 | ||
| AK083494.1 | - | 1 | 54 | |||
| BC082334.1 | - | 3 | 1400 | |||