siRNA Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110421.1 | 6 | 1199 | |||
| NR_110422.1 | 5 | 1073 | ||||
| NR_110423.1 | 4 | 1063 | ||||
| NR_110424.1 | 4 | 1046 | ||||
| NR_110425.1 | 4 | 961 | ||||
| GenBank mRNA | AK013290.1 | - | 4 | 803 | ||
| BC037766.1 | - | 3 | 1391 | |||
| BC054406.1 | - | 4 | 807 | |||

Eldr
Genomic Map