siRNA Details
| Gene Symbol: 2810442N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040562.1 | 2 | 631 | |||
| GenBank mRNA | AK013293.1 | - | 2 | 632 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712295.1 | - | 2 | 632 | Chr. 1 - 162004969 - 162005824 [+] on Build GRCm38 | |

2810442N19Rik
Genomic Map