siRNA Details
| Gene Symbol: 2210414B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040643.1 | 2 | 280 | |||
| GenBank mRNA | AA165904.1 | - | 2 | 278 | ||
| AI509281.1 | - | 2 | 280 | |||
| AK008921.1 | - | 2 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210414B05Rik| Gene Symbol: 2210414B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040643.1 | 2 | 280 | |||
| GenBank mRNA | AA165904.1 | - | 2 | 278 | ||
| AI509281.1 | - | 2 | 280 | |||
| AK008921.1 | - | 2 | 274 | |||