siRNA Details
| Gene Symbol: 2210414I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008926.1 | - | 1 | 468 | ||
| AK132110.1 | - | 1 | 596 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ044771.1 | - | 1 | 596 | Chr. 2 - 164911890 - 164912590 [+] on Build GRCm38 | |

2210414I22Rik
Genomic Map