siRNA Details
| Gene Symbol: 2210414I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008926.1 | - | 1 | 571 | ||
| AK132110.1 | - | 1 | 699 | |||
| AV080469.1 | - | 1 | 94 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210414I22Rik| Gene Symbol: 2210414I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008926.1 | - | 1 | 571 | ||
| AK132110.1 | - | 1 | 699 | |||
| AV080469.1 | - | 1 | 94 | |||